Learn More
Medchemexpress LLC D-Sorbitol-13C6 | 121067-66-1 | 99.94% | 1 MG
SDP
Catalog No. 5000406329
Encompass_Preferred
D-Sorbitol-13C6 is a 13C labeled version of D-Sorbitol, a six-carbon sugar alcohol. It functions as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This compound is also utilized as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Functions as a sugar substitute
  • Acts as a stabilizing excipient and/or isotonicity agent
  • Can be used as a sweetener, humectant, and thickener
  • Serves as a dietary supplement
  • Utilized as a tracer in quantitative analysis
  • Functions as an internal standard in quantitative analysis
Catalog No. 50-004-06329 Supplier Medchemexpress LLC Supplier No. HYB0400S161MG
D-Sorbitol-13C6 is a 13C labeled version of D-Sorbitol, a six-carbon sugar alcohol. It functions as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This compound is also utilized as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Functions as a sugar substitute
  • Acts as a stabilizing excipient and/or isotonicity agent
  • Can be used as a sweetener, humectant, and thickener
  • Serves as a dietary supplement
  • Utilized as a tracer in quantitative analysis
  • Functions as an internal standard in quantitative analysis