Learn More
Medchemexpress LLC D-sorbitol-13C6 | 121067-66-1 | MFCD08459669 | 99.9% | 188.13 | 13C6H14O6 | 10 MG
SDP
Catalog No. 5000298062
Encompass_Preferred
D-Sorbitol-13C6 is the uniformly 13C-labeled isotopologue of D-sorbitol (D-glucitol), supplied for research use as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It has formula 13C6H14O6 and a reported purity of 99.94%, and is offered in small milligram quantities for analytical workflows.
  • Uniformly 13C-labeled D-sorbitol (13C6) suitable as a tracer.
  • High purity suitable for quantitative analytical applications.
  • Used as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in small mg sizes to support method development and calibration.
  • Supplied as a solid isotopically labeled compound for research use only.
Catalog No. 50-002-98062 Supplier Medchemexpress LLC Supplier No. HYB0400S1610MG
D-Sorbitol-13C6 is the uniformly 13C-labeled isotopologue of D-sorbitol (D-glucitol), supplied for research use as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It has formula 13C6H14O6 and a reported purity of 99.94%, and is offered in small milligram quantities for analytical workflows.
  • Uniformly 13C-labeled D-sorbitol (13C6) suitable as a tracer.
  • High purity suitable for quantitative analytical applications.
  • Used as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in small mg sizes to support method development and calibration.
  • Supplied as a solid isotopically labeled compound for research use only.