Learn More
Medchemexpress LLC D-Sorbitol | 50-70-4 | 99.9% | 1 G
SDP
Catalog No. 5000406657
Encompass_Preferred
D-Sorbitol is a six-carbon sugar alcohol that serves as a sugar substitute. It is produced from glucose or sucrose by hydrogenation or by bacteria through an enzymatic process. This compound functions as a versatile excipient and additive in various applications.
  • Functions as a sugar substitute
  • Can be used as a stabilizing excipient and isotonicity agent
  • Serves as a sweetener, humectant, and thickener
  • Acts as a dietary supplement
  • Used as a fast disintegrant in capsules
  • Plasticizer in capsule shells and tablet film coatings
  • Drug stabilizer in oral liquids
  • Solubility enhancer for drugs like indomethacin
  • Humectant in topical formulations
Catalog No. 50-004-06657 Supplier Medchemexpress LLC Supplier No. HYB04001G
D-Sorbitol is a six-carbon sugar alcohol that serves as a sugar substitute. It is produced from glucose or sucrose by hydrogenation or by bacteria through an enzymatic process. This compound functions as a versatile excipient and additive in various applications.
  • Functions as a sugar substitute
  • Can be used as a stabilizing excipient and isotonicity agent
  • Serves as a sweetener, humectant, and thickener
  • Acts as a dietary supplement
  • Used as a fast disintegrant in capsules
  • Plasticizer in capsule shells and tablet film coatings
  • Drug stabilizer in oral liquids
  • Solubility enhancer for drugs like indomethacin
  • Humectant in topical formulations