Learn More
Medchemexpress LLC D-Sorbitol | 50-70-4 | 99.95% | 5 G
Supplier: Medchemexpress LLC HYB04005G
D-Sorbitol is a six-carbon sugar alcohol that can be used as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement.
- Functions as a fast disintegrant in capsules.
- Acts as a plasticizer in capsule shells and tablet film coatings.
- Serves as a sugar substitute and drug stabilizer in oral liquids.
- Enhances the solubility of drugs such as indomethacin.
- Used as a stabilizing excipient and/or isotonicity agent in liquid and lyophilized parenteral protein formulations.
- Employed as a humectant in topical formulations.