Learn More
Apexbio Technology LLC D-Sorbitol 98201-93-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094119
Encompass_Preferred
D-Sorbitol is a polyhydric sugar alcohol widely used as an osmotic stabilizer in biological and pharmaceutical research. D-Sorbitol exerts its biological activity primarily through inducing hyperosmotic stress conditions at the cellular level, thereby facilitating the study of osmoregulation mechanisms, stress-responsive signaling pathways, and protein stabilization strategies. As a chemical reagent, it serves as a substrate to analyze enzyme kinetics involving aldose reductase and sorbitol dehydrogenase in metabolic research protocols. Based on these properties, D-Sorbitol holds research potential in cell isolation, tissue culture, studies of the polyol pathway, diabetic complications, and energy balance assays.
Catalog No. 50-000-94119 Supplier Apexbio Technology LLC Supplier No. B33245.1
D-Sorbitol is a polyhydric sugar alcohol widely used as an osmotic stabilizer in biological and pharmaceutical research. D-Sorbitol exerts its biological activity primarily through inducing hyperosmotic stress conditions at the cellular level, thereby facilitating the study of osmoregulation mechanisms, stress-responsive signaling pathways, and protein stabilization strategies. As a chemical reagent, it serves as a substrate to analyze enzyme kinetics involving aldose reductase and sorbitol dehydrogenase in metabolic research protocols. Based on these properties, D-Sorbitol holds research potential in cell isolation, tissue culture, studies of the polyol pathway, diabetic complications, and energy balance assays.