Learn More
Medchemexpress LLC D-Sorbitol, for cell culture | 50-70-4 | 98.0% | 25 G
SDP
Catalog No. 5000407076
Encompass_Preferred
D-Sorbitol, also known as D-Glucitol, is a six-carbon sugar alcohol primarily used as a biochemical reagent in life science research. It can function as a biological material or organic compound, and also serves as a sugar substitute and various pharmaceutical excipient.
  • Used as a biological material or organic compound for life science research.
  • Functions as a stabilizing excipient and isotonicity agent.
  • Can be utilized as a fast disintegrant in capsules.
  • Acts as a plasticizer in capsule shells and tablet film coatings.
  • Serves as a sugar substitute and drug stabilizer in oral liquids.
  • Enhances solubility for various drugs.
  • Employed as a humectant in topical formulations.
Catalog No. 50-004-07076 Supplier Medchemexpress LLC Supplier No. 25GHYB0400B
D-Sorbitol, also known as D-Glucitol, is a six-carbon sugar alcohol primarily used as a biochemical reagent in life science research. It can function as a biological material or organic compound, and also serves as a sugar substitute and various pharmaceutical excipient.
  • Used as a biological material or organic compound for life science research.
  • Functions as a stabilizing excipient and isotonicity agent.
  • Can be utilized as a fast disintegrant in capsules.
  • Acts as a plasticizer in capsule shells and tablet film coatings.
  • Serves as a sugar substitute and drug stabilizer in oral liquids.
  • Enhances solubility for various drugs.
  • Employed as a humectant in topical formulations.