Learn More
Medchemexpress LLC D-Sorbitol, for cell culture | 50-70-4 | 98.0% | 250 G
SDP
Catalog No. 5000405602
Encompass_Preferred
D-Sorbitol, also known as D-Glucitol, is a biochemical reagent primarily used for cell culture applications. This six-carbon sugar alcohol can be utilized as a biological material or an organic compound for life science-related research. It functions as a sugar substitute and stabilizing excipient.
  • Used as a biological material or organic compound for life science research.
  • Functions as a sugar substitute.
  • Serves as a stabilizing excipient and/or isotonicity agent.
  • Acts as a sweetener, humectant, or thickener.
  • Can be a dietary supplement.
  • Used as a fast disintegrant in capsules.
  • Plasticizer in capsule shells and tablet film coatings.
  • Drug stabilizer in oral liquids.
  • Solubility enhancer for drugs.
  • Used in topical formulations as a humectant.
Catalog No. 50-004-05602 Supplier Medchemexpress LLC Supplier No. 250GHYB0400B
D-Sorbitol, also known as D-Glucitol, is a biochemical reagent primarily used for cell culture applications. This six-carbon sugar alcohol can be utilized as a biological material or an organic compound for life science-related research. It functions as a sugar substitute and stabilizing excipient.
  • Used as a biological material or organic compound for life science research.
  • Functions as a sugar substitute.
  • Serves as a stabilizing excipient and/or isotonicity agent.
  • Acts as a sweetener, humectant, or thickener.
  • Can be a dietary supplement.
  • Used as a fast disintegrant in capsules.
  • Plasticizer in capsule shells and tablet film coatings.
  • Drug stabilizer in oral liquids.
  • Solubility enhancer for drugs.
  • Used in topical formulations as a humectant.