Learn More
Medchemexpress LLC D-Sorbitol-d8 | 287962-59-8 | 98.9% | 1 MG
SDP
Catalog No. 5000407328
Encompass_Preferred
D-Sorbitol-d8 is a deuterium-labeled version of D-Sorbitol, a six-carbon sugar alcohol. D-Sorbitol can be used as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This product is intended for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Functions as a stabilizing excipient
  • Acts as an isotonicity agent
  • Serves as a sweetener
  • Used as a humectant
  • Functions as a thickener
  • Can be used as a dietary supplement
Catalog No. 50-004-07328 Supplier Medchemexpress LLC Supplier No. HYB0400S1MG
D-Sorbitol-d8 is a deuterium-labeled version of D-Sorbitol, a six-carbon sugar alcohol. D-Sorbitol can be used as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This product is intended for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Functions as a stabilizing excipient
  • Acts as an isotonicity agent
  • Serves as a sweetener
  • Used as a humectant
  • Functions as a thickener
  • Can be used as a dietary supplement