Learn More
Medchemexpress LLC D-Sorbitol-d8 | 287962-59-8 | 98.9% | 5 MG
SDP
Catalog No. 5000406104
Encompass_Preferred
This compound is a deuterium labeled six-carbon sugar alcohol, which can be utilized as a sugar substitute. It functions as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06104 Supplier Medchemexpress LLC Supplier No. HYB0400S5MG
This compound is a deuterium labeled six-carbon sugar alcohol, which can be utilized as a sugar substitute. It functions as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.