Learn More
Medchemexpress LLC D-Sorbitol (Standard) | 50-70-4 | >98.7% | 50 MG
SDP
Catalog No. 5000407152
Encompass_Preferred
D-Sorbitol (Standard) is the analytical standard for D-Sorbitol, a six-carbon sugar alcohol. It serves as a sugar substitute and finds use in research and analytical applications. It can function as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This product is designated for research use only.
  • The compound is an analytical standard, providing a reference standard with a supplied assay.
  • It is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • The product is intended for research and analytical applications.
  • It can be used as a sugar substitute.
  • It can act as a stabilizing excipient and/or isotonicity agent.
  • It functions as a sweetener and humectant.
  • It can be used as a thickener and dietary supplement.
Catalog No. 50-004-07152 Supplier Medchemexpress LLC Supplier No. HYB0400R50MG
D-Sorbitol (Standard) is the analytical standard for D-Sorbitol, a six-carbon sugar alcohol. It serves as a sugar substitute and finds use in research and analytical applications. It can function as a stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This product is designated for research use only.
  • The compound is an analytical standard, providing a reference standard with a supplied assay.
  • It is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • The product is intended for research and analytical applications.
  • It can be used as a sugar substitute.
  • It can act as a stabilizing excipient and/or isotonicity agent.
  • It functions as a sweetener and humectant.
  • It can be used as a thickener and dietary supplement.