Learn More
Medchemexpress LLC D-Sorbitol (Standard) | 50-70-4 | 98.7% | 500 MG
SDP
Catalog No. 5000406559
Encompass_Preferred
D-Sorbitol (Standard) is the analytical standard of D-Sorbitol, a six-carbon sugar alcohol used in research and analytical applications. It functions as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This analytical grade compound serves as a reference standard, commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for D-Sorbitol
  • Six-carbon sugar alcohol
  • Can be used as a sugar substitute
  • Functions as a stabilizing excipient and isotonicity agent
  • Also acts as a sweetener, humectant, thickener, and dietary supplement
  • Ideal for qualitative, quantitative, and methodological research (HPLC, GC, MS)
  • Appears as a white to off-white solid
  • Assay purity: 98.67%
Catalog No. 50-004-06559 Supplier Medchemexpress LLC Supplier No. HYB0400R500MG
D-Sorbitol (Standard) is the analytical standard of D-Sorbitol, a six-carbon sugar alcohol used in research and analytical applications. It functions as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This analytical grade compound serves as a reference standard, commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for D-Sorbitol
  • Six-carbon sugar alcohol
  • Can be used as a sugar substitute
  • Functions as a stabilizing excipient and isotonicity agent
  • Also acts as a sweetener, humectant, thickener, and dietary supplement
  • Ideal for qualitative, quantitative, and methodological research (HPLC, GC, MS)
  • Appears as a white to off-white solid
  • Assay purity: 98.67%