Learn More
Medchemexpress LLC D-Sorbitol (Standard) | 50-70-4 | MFCD00004708 | 98.7% | 250 MG
SDP
Catalog No. 5000406806
Encompass_Preferred
D-Sorbitol (Standard) is the analytical standard of D-Sorbitol. It is a six-carbon sugar alcohol and can be used as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This product is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for D-Sorbitol
  • Six-carbon sugar alcohol
  • Can be used as a sugar substitute
  • Acts as a stabilizing excipient and/or isotonicity agent
  • Functions as a sweetener, humectant, and thickener
  • Suitable for dietary supplements
  • Intended for research and analytical applications
  • Commonly used in HPLC, GC, and MS experiments
Catalog No. 50-004-06806 Supplier Medchemexpress LLC Supplier No. 250MGHYB0400R
D-Sorbitol (Standard) is the analytical standard of D-Sorbitol. It is a six-carbon sugar alcohol and can be used as a sugar substitute, stabilizing excipient, isotonicity agent, sweetener, humectant, thickener, and dietary supplement. This product is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for D-Sorbitol
  • Six-carbon sugar alcohol
  • Can be used as a sugar substitute
  • Acts as a stabilizing excipient and/or isotonicity agent
  • Functions as a sweetener, humectant, and thickener
  • Suitable for dietary supplements
  • Intended for research and analytical applications
  • Commonly used in HPLC, GC, and MS experiments