Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(+)-sorbose | 3615-56-3 | MFCD00151095 | 99.2% | 180.16 g/mol | C6H12O6 | 10 MG
SDP

Catalog No. 5000283721
Encompass_Preferred

D-(+)-Sorbose is the D-enantiomer of sorbose supplied for research use. It is a water-soluble monosaccharide sweetener used in metabolic and enzymatic studies and has been reported to inhibit disaccharidase activity and suppress postprandial blood glucose and insulin in animal models. Not for human consumption.

  • High purity: 99.2%.
  • Molecular weight: 180.16 g/mol.
  • Chemical formula: C6H12O6.
  • Soluble in water: 125 mg/mL (requires ultrasonic).
  • Storage: store at 4°C and protect from light.

Catalog No. 50-002-83721 Supplier Medchemexpress LLC Supplier No. HYW14549710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Sorbose is the D-enantiomer of sorbose supplied for research use. It is a water-soluble monosaccharide sweetener used in metabolic and enzymatic studies and has been reported to inhibit disaccharidase activity and suppress postprandial blood glucose and insulin in animal models. Not for human consumption.

  • High purity: 99.2%.
  • Molecular weight: 180.16 g/mol.
  • Chemical formula: C6H12O6.
  • Soluble in water: 125 mg/mL (requires ultrasonic).
  • Storage: store at 4°C and protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.