Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Sorbose 98.0+%, TCI America™
SDP

Catalog No. S0390100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0390100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0390100MG Supplier TCI America Supplier No. S0390100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3615-56-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00151095
InChI Key LKDRXBCSQODPBY-IANNHFEVSA-N
Synonym d-sorbose, keto-d-sorbose, unii-jj09461njs, 3r,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, xylo-hexulose, sorbose, d, d-sor, d-xylo-hex-2-ulose, d-+-sorbose
PubChem CID 107428
ChEBI CHEBI:13022
IUPAC Name (3R,4S,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OCC1(O)OC[C@@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 158°C
Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-Sorbose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.