Learn More
Medchemexpress LLC D-streptamine, O-2-amino-2,3,4,6-tetradeoxy-6-(methylamino)-alpha-D-erythro-hexopyranosyl sulfate | 66803-19-8 | 100.0% | 463.6 g/mol | C20H41N5O7.xH2SO4 | 100 MG
SDP
Catalog No. 5000242643
Encompass_Preferred
Micronomicin sulfate is an aminoglycoside antibiotic supplied as the sulfate salt for research use. It is isolated from Micromonospora and exhibits broad-spectrum activity against Gram-negative bacteria. The product is provided as a research-grade powder with manufacturer-available technical documents including a datasheet, certificate of analysis, and safety data sheet.
  • High reported purity (99.96%).
  • Water-soluble powder suitable for solution preparation.
  • Broad-spectrum antibacterial activity useful for in vitro studies.
  • Provided with datasheet, certificate of analysis, and safety data sheet for traceability.
  • Suitable for microbiology and antibiotic research applications.
Catalog No. 50-002-42643 Supplier Medchemexpress LLC Supplier No. HY108307100MG
Micronomicin sulfate is an aminoglycoside antibiotic supplied as the sulfate salt for research use. It is isolated from Micromonospora and exhibits broad-spectrum activity against Gram-negative bacteria. The product is provided as a research-grade powder with manufacturer-available technical documents including a datasheet, certificate of analysis, and safety data sheet.
  • High reported purity (99.96%).
  • Water-soluble powder suitable for solution preparation.
  • Broad-spectrum antibacterial activity useful for in vitro studies.
  • Provided with datasheet, certificate of analysis, and safety data sheet for traceability.
  • Suitable for microbiology and antibiotic research applications.