Learn More
Medchemexpress LLC D-(+)-Talose-13C-1 | 83379-36-6 | 99.0% | 181.15 g/mol | C5(13C)H12O6 | 5 MG

Supplier: Medchemexpress LLC HYW145492S35MG
D-(+)-Talose-13C-1 is a stable 13C-labeled monosaccharide provided for analytical and biochemical research. It is intended for use as a tracer or internal standard in metabolic tracing and analytical workflows.
- Labeled with 13C at carbon 1 for isotope tracing.
- High purity: 99.0%.
- Suitable as a tracer and internal standard for mass spectrometry and NMR analyses.
- Supplied in small research quantities for analytical workflows.
- Molecular weight 181.15 g/mol; formula C5(13C)H12O6.
- CAS number 83379-36-6 for substance identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.