Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(-)-tartaric acid diisopropyl ester | 62961-64-2 | MFCD00008876 | 234.25 g/mol | C10H18O6 | 100 G
SDP

Catalog No. 5000300506
Encompass_Preferred

D-(-)-Tartaric acid diisopropyl ester is a chiral organic reagent used as a chemical building block and biochemical reagent in life-science research. It is the diisopropyl ester derivative of D-tartaric acid and is supplied for use in synthesis, enantioselective transformations, and analytical applications.

  • Chiral diisopropyl ester of D-tartaric acid for enantioselective synthesis.
  • Suitable as a reagent and building block in organic synthesis and biochemical assays.
  • Molecular formula C10H18O6; molecular weight 234.25 g/mol.
  • Available in multiple package sizes from 10 g to 500 g.
  • Storage recommendations: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Product documentation includes a data sheet and safety data sheet.

Catalog No. 50-003-00506 Supplier Medchemexpress LLC Supplier No. HYW037483100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(-)-Tartaric acid diisopropyl ester is a chiral organic reagent used as a chemical building block and biochemical reagent in life-science research. It is the diisopropyl ester derivative of D-tartaric acid and is supplied for use in synthesis, enantioselective transformations, and analytical applications.

  • Chiral diisopropyl ester of D-tartaric acid for enantioselective synthesis.
  • Suitable as a reagent and building block in organic synthesis and biochemical assays.
  • Molecular formula C10H18O6; molecular weight 234.25 g/mol.
  • Available in multiple package sizes from 10 g to 500 g.
  • Storage recommendations: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Product documentation includes a data sheet and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.