Learn More
Medchemexpress LLC D-(-)-tartaric acid diisopropyl ester | 62961-64-2 | MFCD00008876 | >98.0% | 234.25 g/mol | C10H18O6 | 10 G
SDP
Catalog No. 5000300367
Encompass_Preferred
D-(-)-Tartaric acid diisopropyl ester is the diisopropyl ester derivative of D-tartaric acid used as a chiral building block and biochemical reagent for research applications. It is supplied as a colorless to light yellow liquid (MW 234.25 g/mol) and is commonly used in enantioselective synthesis and chiral derivatization workflows.
  • Chiral building block for asymmetric synthesis.
  • Useful for chiral derivatization and analytical methods.
  • Liquid form facilitates solution-phase reactions and handling.
  • Compatible with common organic solvents.
  • Storage-stable when kept under recommended temperatures.
Catalog No. 50-003-00367 Supplier Medchemexpress LLC Supplier No. HYW03748310G
D-(-)-Tartaric acid diisopropyl ester is the diisopropyl ester derivative of D-tartaric acid used as a chiral building block and biochemical reagent for research applications. It is supplied as a colorless to light yellow liquid (MW 234.25 g/mol) and is commonly used in enantioselective synthesis and chiral derivatization workflows.
  • Chiral building block for asymmetric synthesis.
  • Useful for chiral derivatization and analytical methods.
  • Liquid form facilitates solution-phase reactions and handling.
  • Compatible with common organic solvents.
  • Storage-stable when kept under recommended temperatures.