Learn More
Medchemexpress LLC D-(-)-tartaric acid diisopropyl ester | 62961-64-2 | MFCD00008876 | >98.0% | 234.25 g/mol | C10H18O6 | 500 G
SDP
Catalog No. 5000301895
Encompass_Preferred
D-(-)-Tartaric acid diisopropyl ester is a chiral organic reagent used in synthetic and analytical chemistry for enantioselective reactions, resolution of racemates, and as a chiral building block. It is supplied as a colorless to light yellow liquid with a listed density of 1.119 g/cm3 and is typically handled and stored under cold conditions for long-term stability.
  • Chiral building block for enantioselective synthesis.
  • Suitable for kinetic resolution and chiral auxiliary applications.
  • Colorless to light yellow liquid with density 1.119 g/cm3.
  • Typical storage recommendations include cold conditions for long-term stability.
  • Available in multiple laboratory pack sizes up to 500 g.
  • Purity commonly reported as >98.0% by GC (supplier-specific).
Catalog No. 50-003-01895 Supplier Medchemexpress LLC Supplier No. HYW037483500G
D-(-)-Tartaric acid diisopropyl ester is a chiral organic reagent used in synthetic and analytical chemistry for enantioselective reactions, resolution of racemates, and as a chiral building block. It is supplied as a colorless to light yellow liquid with a listed density of 1.119 g/cm3 and is typically handled and stored under cold conditions for long-term stability.
  • Chiral building block for enantioselective synthesis.
  • Suitable for kinetic resolution and chiral auxiliary applications.
  • Colorless to light yellow liquid with density 1.119 g/cm3.
  • Typical storage recommendations include cold conditions for long-term stability.
  • Available in multiple laboratory pack sizes up to 500 g.
  • Purity commonly reported as >98.0% by GC (supplier-specific).