Learn More
Cayman Chemical D-TetrahydropalmatIn 50mg
SDP
Catalog No. 502913354
Encompass_Preferred
A tetrahydroberberine alkaloid; reverses apomorphine-induced inhibition of dopaminergic firing in the SNC in paralyzed rats at 32 mg/kg
Catalog No. 50-291-3354 Supplier Cayman Chemical Supplier No. 3557750 MG
A tetrahydroberberine alkaloid; reverses apomorphine-induced inhibition of dopaminergic firing in the SNC in paralyzed rats at 32 mg/kg