Learn More
Medchemexpress LLC D-Tetrahydropalmatine | 3520-14-7 | 355.43 | 5 MG
SDP
Catalog No. 5000416645
Encompass_Preferred
D-Tetrahydropalmatine is an isoquinoline alkaloid found primarily in the genus Corydalis. It functions as a dopamine (DA) receptor antagonist with preferential affinity for D1 receptors, and also acts as a potent inhibitor of organic cation transporter 1 (OCT1).
  • Dopamine D1 receptor antagonist
  • Potent organic cation transporter 1 (OCT1) inhibitor
  • Isoquinoline alkaloid
  • Derived from plants
Catalog No. 50-004-16645 Supplier Medchemexpress LLC Supplier No. HYN20035MG
D-Tetrahydropalmatine is an isoquinoline alkaloid found primarily in the genus Corydalis. It functions as a dopamine (DA) receptor antagonist with preferential affinity for D1 receptors, and also acts as a potent inhibitor of organic cation transporter 1 (OCT1).
  • Dopamine D1 receptor antagonist
  • Potent organic cation transporter 1 (OCT1) inhibitor
  • Isoquinoline alkaloid
  • Derived from plants