Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Tetrahydropalmatine | 3520-14-7 | MFCD11046348 | 355.43 | 1 ML
SDP

Catalog No. 5000414611
Encompass_Preferred

D-Tetrahydropalmatine is an isoquinoline alkaloid primarily found in the genus Corydalis. It acts as a dopamine (DA) receptor antagonist, showing preferential affinity for D1 receptors. Additionally, it is identified as a potent inhibitor of organic cation transporter 1 (OCT1).

  • Dopamine receptor antagonist
  • Preferential affinity toward the D1 receptors
  • Potent organic cation transporter 1 (OCT1) inhibitor
  • Purity: 99.84%
  • Appearance: Solid
  • Color: White to light yellow
  • Initial source: Plants Papaveraceae Corydalis yanhusuo W. T. Wang

Catalog No. 50-004-14611 Supplier Medchemexpress LLC Supplier No. HYN200310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Tetrahydropalmatine is an isoquinoline alkaloid primarily found in the genus Corydalis. It acts as a dopamine (DA) receptor antagonist, showing preferential affinity for D1 receptors. Additionally, it is identified as a potent inhibitor of organic cation transporter 1 (OCT1).

  • Dopamine receptor antagonist
  • Preferential affinity toward the D1 receptors
  • Potent organic cation transporter 1 (OCT1) inhibitor
  • Purity: 99.84%
  • Appearance: Solid
  • Color: White to light yellow
  • Initial source: Plants Papaveraceae Corydalis yanhusuo W. T. Wang

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.