Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-tetrahydropalmatine | 3520-14-7 | MFCD11046348 | 99.8% | 355.43 | C21H25NO4 | 10 MG
SDP

Catalog No. 5000296147
Encompass_Preferred

D-Tetrahydropalmatine is an isoquinoline alkaloid research reagent that acts as a dopamine receptor antagonist with preferential affinity for D1 receptors and reported inhibition of organic cation transporter 1 (OCT1). It is supplied as a solid for pharmacological and biochemical studies of dopamine signaling.

  • CAS number: 3520-14-7.
  • Molecular formula: C21H25NO4.
  • Molecular weight: 355.43.
  • Purity: 99.8%.
  • Appearance: white to light yellow solid.
  • Solubility: 50 mg/mL in DMSO (≈140.67 mM).
  • Storage: powder at -20°C for up to 3 years; in solution at -80°C for extended storage.
  • Intended use: for research only, not for diagnostic or therapeutic applications.

Catalog No. 50-002-96147 Supplier Medchemexpress LLC Supplier No. HYN200310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Tetrahydropalmatine is an isoquinoline alkaloid research reagent that acts as a dopamine receptor antagonist with preferential affinity for D1 receptors and reported inhibition of organic cation transporter 1 (OCT1). It is supplied as a solid for pharmacological and biochemical studies of dopamine signaling.

  • CAS number: 3520-14-7.
  • Molecular formula: C21H25NO4.
  • Molecular weight: 355.43.
  • Purity: 99.8%.
  • Appearance: white to light yellow solid.
  • Solubility: 50 mg/mL in DMSO (≈140.67 mM).
  • Storage: powder at -20°C for up to 3 years; in solution at -80°C for extended storage.
  • Intended use: for research only, not for diagnostic or therapeutic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.