Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Threitol | 2418-52-2 | 99.95% | 122.12 | 10 MG
SDP

Catalog No. 5000454874
Encompass_Preferred

D-threitol serves as an antifreeze agent in the Alaskan beetle *Upis ceramboides*.

  • Purity: 99.95%
  • Molecular weight: 122.12
  • Formula: C4H10O4
  • CAS number: 2418-52-2
  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Saccharides
  • Storage (powder): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Solubility in DMSO: 250 mg/mL (2047.17 mM)
  • Initial source: Endogenous metabolite, disease markers, endocrine diseases, microorganisms

Catalog No. 50-004-54874 Supplier Medchemexpress LLC Supplier No. HYW01284610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

D-threitol serves as an antifreeze agent in the Alaskan beetle *Upis ceramboides*.

  • Purity: 99.95%
  • Molecular weight: 122.12
  • Formula: C4H10O4
  • CAS number: 2418-52-2
  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Saccharides
  • Storage (powder): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Solubility in DMSO: 250 mg/mL (2047.17 mM)
  • Initial source: Endogenous metabolite, disease markers, endocrine diseases, microorganisms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.