Learn More
Cayman Chemical D-threo-Biopterin, Purity: ≥98%, CAS: 13019-52-8, 5 mg
SDP
Catalog No. 502991647
Encompass_Preferred
A pterin; enhances luminol chemiluminescence induced by the oxidant chloramine-T in a cell-free assay at 100 μM
Catalog No. 50-299-1647 Supplier Cayman Chemical Supplier No. 299865 MG
A pterin; enhances luminol chemiluminescence induced by the oxidant chloramine-T in a cell-free assay at 100 μM