Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-threo-PDMP 25mg | 109836-82-0 | 390.56 | C23H38N2O3 | 25 MG
SDP

Catalog No. 5000227348
Encompass_Preferred

D-threo-PDMP is a small-molecule research compound that inhibits glucosylceramide synthase (GCS) and is used to modulate glycosphingolipid synthesis in biochemical and cellular studies. It is supplied as an oil and offered in multiple vial sizes, including a 25 mg quantity.

  • Glucosylceramide synthase inhibitor used in cellular and biochemical assays.
  • CAS number 109836-82-0.
  • Molecular formula C23H38N2O3; molecular weight 390.56 Da.
  • High purity (99.5%) suitable for research applications.
  • Supplied as an oil; available vial sizes include 5 mg-500 mg and a 10 mM solution in DMSO.
  • Used to study apoptosis, cell adhesion, and glycosphingolipid metabolism.

Catalog No. 50-002-27348 Supplier Medchemexpress LLC Supplier No. HY116392E25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-threo-PDMP is a small-molecule research compound that inhibits glucosylceramide synthase (GCS) and is used to modulate glycosphingolipid synthesis in biochemical and cellular studies. It is supplied as an oil and offered in multiple vial sizes, including a 25 mg quantity.

  • Glucosylceramide synthase inhibitor used in cellular and biochemical assays.
  • CAS number 109836-82-0.
  • Molecular formula C23H38N2O3; molecular weight 390.56 Da.
  • High purity (99.5%) suitable for research applications.
  • Supplied as an oil; available vial sizes include 5 mg-500 mg and a 10 mM solution in DMSO.
  • Used to study apoptosis, cell adhesion, and glycosphingolipid metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.