Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-threo-PDMP hydroch 1mg | 139889-62-6 | 1 MG
SDP

Catalog No. 5000229875
Encompass_Preferred

D-threo-PDMP hydrochloride is a research compound that inhibits glucosylceramide synthase (GCS). It reduces cellular glycosphingolipids such as GM3 and GD3 and is used to study sphingolipid metabolism and related cellular processes including neurite growth, cell adhesion, and apoptosis.

  • Potent glucosylceramide synthase inhibitor used in cellular studies.
  • High purity (98.0%) suitable for research applications.
  • White to off-white solid, supplied in milligram quantities for laboratory use.
  • Stable when stored sealed at -20 °C; solutions have recommended storage conditions.
  • Molecular weight 427.02; molecular formula C23H39ClN2O3.

Catalog No. 50-002-29875 Supplier Medchemexpress LLC Supplier No. HY116392F1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-threo-PDMP hydrochloride is a research compound that inhibits glucosylceramide synthase (GCS). It reduces cellular glycosphingolipids such as GM3 and GD3 and is used to study sphingolipid metabolism and related cellular processes including neurite growth, cell adhesion, and apoptosis.

  • Potent glucosylceramide synthase inhibitor used in cellular studies.
  • High purity (98.0%) suitable for research applications.
  • White to off-white solid, supplied in milligram quantities for laboratory use.
  • Stable when stored sealed at -20 °C; solutions have recommended storage conditions.
  • Molecular weight 427.02; molecular formula C23H39ClN2O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.