Learn More
Medchemexpress LLC D-threo-PDMP hydroch 5mg | 139889-62-6 | 427.02 g/mol | C23H39ClN2O3 | 5 MG
SDP
Catalog No. 5000227158
Encompass_Preferred
D-threo-PDMP hydrochloride is the hydrochloride salt of a potent glucosylceramide synthase (GCS) inhibitor used in biochemical and cellular research to study glycosphingolipid metabolism. It is supplied as a white to off-white solid with high purity for in vitro applications.
  • High purity: 98.0%.
  • Hydrochloride salt for improved handling and solubility.
  • White to off-white solid appearance.
  • Soluble in ethanol and methanol for preparation of stock solutions.
  • Suitable for in vitro studies of glycosphingolipid metabolism and GCS inhibition.
  • Supplied in a 5 MG quantity for small-scale research use.
Catalog No. 50-002-27158 Supplier Medchemexpress LLC Supplier No. HY116392F5MG
D-threo-PDMP hydrochloride is the hydrochloride salt of a potent glucosylceramide synthase (GCS) inhibitor used in biochemical and cellular research to study glycosphingolipid metabolism. It is supplied as a white to off-white solid with high purity for in vitro applications.
  • High purity: 98.0%.
  • Hydrochloride salt for improved handling and solubility.
  • White to off-white solid appearance.
  • Soluble in ethanol and methanol for preparation of stock solutions.
  • Suitable for in vitro studies of glycosphingolipid metabolism and GCS inhibition.
  • Supplied in a 5 MG quantity for small-scale research use.