Learn More
Medchemexpress LLC D-threo-PPMP hydroch 1mg | 139889-65-9 | 1 MG
SDP
Catalog No. 5000227375
Encompass_Preferred
D-threo-PPMP hydrochloride is a research-grade small molecule inhibitor of glucosylceramide (GlcCer) synthase used in cell biology to block karyokinesis and reduce cyst production.
  • Potent inhibitor of glucosylceramide (GlcCer) synthase.
  • Active enantiomer used in cellular studies to block karyokinesis.
  • Solid form with >98.0% purity.
  • Molecular weight 511.18 Da and formula C29H51ClN2O3.
  • Recommended sealed storage at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-27375 Supplier Medchemexpress LLC Supplier No. HY1165351MG
D-threo-PPMP hydrochloride is a research-grade small molecule inhibitor of glucosylceramide (GlcCer) synthase used in cell biology to block karyokinesis and reduce cyst production.
  • Potent inhibitor of glucosylceramide (GlcCer) synthase.
  • Active enantiomer used in cellular studies to block karyokinesis.
  • Solid form with >98.0% purity.
  • Molecular weight 511.18 Da and formula C29H51ClN2O3.
  • Recommended sealed storage at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).