Learn More
Medchemexpress LLC D-threo-PPMP hydroch 5mg | 139889-65-9 | 5 MG
SDP
Catalog No. 5000308268
Encompass_Preferred
D-threo-PPMP hydrochloride is the hydrochloride salt of D-threo-PPMP, a potent inhibitor of glucosylceramide (GlcCer) synthase used in biochemical and cell biology research. It inhibits GlcCer synthesis, and has been reported to block karyokinesis and reduce cyst production.
  • Potent inhibitor of glucosylceramide (GlcCer) synthase.
  • Useful for studies of sphingolipid metabolism and glycosphingolipid biosynthesis.
  • Reported to block karyokinesis and reduce cyst production.
  • High purity (≈98.0%) suitable for research applications.
  • Solid, white to off-white; supplied as small, preweighed vials for convenient handling.
  • Recommended storage: store at -20°C; in solution store at -80°C for long-term stability.
Catalog No. 50-003-08268 Supplier Medchemexpress LLC Supplier No. HY1165355MG
D-threo-PPMP hydrochloride is the hydrochloride salt of D-threo-PPMP, a potent inhibitor of glucosylceramide (GlcCer) synthase used in biochemical and cell biology research. It inhibits GlcCer synthesis, and has been reported to block karyokinesis and reduce cyst production.
  • Potent inhibitor of glucosylceramide (GlcCer) synthase.
  • Useful for studies of sphingolipid metabolism and glycosphingolipid biosynthesis.
  • Reported to block karyokinesis and reduce cyst production.
  • High purity (≈98.0%) suitable for research applications.
  • Solid, white to off-white; supplied as small, preweighed vials for convenient handling.
  • Recommended storage: store at -20°C; in solution store at -80°C for long-term stability.