Learn More
Medchemexpress LLC HY-W012874 500mg Medchemexpress, D-Threonine CAS:632-20-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW012874500MG
Encompass_Preferred
Medchemexpress, HY-W012874 500mg D-Threonine CAS:632-20-2 D-Threonine (H-D-Thr-OH) is an enantiomer of L-threonine. D-Threonine is a metabolite of Saccharomyces cerevisiae. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3340