Learn More
Medchemexpress LLC D-Thyroxine | 51-49-0 | 98.0% | C15H11I4NO4 | 25 MG
SDP
Catalog No. 5000359053
Encompass_Preferred
D-Thyroxine (D-T4) is an orally active thyroid hormone that inhibits the secretion of TSH. It can inhibit goiter, promote metamorphosis of tadpoles, and influence cholesterol metabolism. D-Thyroxine can be used for the study of hypercholesterolemia. This product is intended for research applications only.
  • Orally active thyroid hormone
  • Inhibits the secretion of TSH
  • Can inhibit goiter
  • Promotes metamorphosis of tadpoles
  • Influences cholesterol metabolism
  • Used for the study of hypercholesterolemia
Catalog No. 50-003-59053 Supplier Medchemexpress LLC Supplier No. 25MGHYA0152
D-Thyroxine (D-T4) is an orally active thyroid hormone that inhibits the secretion of TSH. It can inhibit goiter, promote metamorphosis of tadpoles, and influence cholesterol metabolism. D-Thyroxine can be used for the study of hypercholesterolemia. This product is intended for research applications only.
  • Orally active thyroid hormone
  • Inhibits the secretion of TSH
  • Can inhibit goiter
  • Promotes metamorphosis of tadpoles
  • Influences cholesterol metabolism
  • Used for the study of hypercholesterolemia