Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Thyroxine | 51-49-0 | 98.02% | C15H11I4NO4 | 5 MG
SDP

Catalog No. 5000432816
Encompass_Preferred

D-Thyroxine (D-T4) is an orally active thyroid hormone that inhibits the secretion of TSH. It can inhibit goiter, promote metamorphosis of tadpoles, and influence cholesterol metabolism. It is used for the study of hypercholesterolemia.

  • Orally active thyroid hormone
  • Inhibits TSH secretion
  • Can inhibit goiter
  • Promotes metamorphosis of tadpoles
  • Influences cholesterol metabolism
  • Used for the study of hypercholesterolemia
  • Molecular weight: 776.87
  • Chemical formula: C15H11I4NO4
  • CAS number: 51-49-0
  • Solid form
  • Light yellow to light brown color

Catalog No. 50-004-32816 Supplier Medchemexpress LLC Supplier No. HYA01525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Thyroxine (D-T4) is an orally active thyroid hormone that inhibits the secretion of TSH. It can inhibit goiter, promote metamorphosis of tadpoles, and influence cholesterol metabolism. It is used for the study of hypercholesterolemia.

  • Orally active thyroid hormone
  • Inhibits TSH secretion
  • Can inhibit goiter
  • Promotes metamorphosis of tadpoles
  • Influences cholesterol metabolism
  • Used for the study of hypercholesterolemia
  • Molecular weight: 776.87
  • Chemical formula: C15H11I4NO4
  • CAS number: 51-49-0
  • Solid form
  • Light yellow to light brown color

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.