Learn More
Medchemexpress LLC D-α-Tocopherol acetate | 58-95-7 | 98.3% | 25 G
SDP
Catalog No. 5000414476
Encompass_Preferred
D-α-Tocopherol acetate can be hydrolyzed to d-alpha-tocopherol (VE) and absorbed in the small intestine. This product is intended for research use only.
  • Inhibits LPS-induced production of NO, prostaglandin E2 and pro-inflammatory cytokines
  • Exhibits slight suppression on the activation of NF-κB p50 and p65
Catalog No. 50-004-14476 Supplier Medchemexpress LLC Supplier No. 25GHYB1278
D-α-Tocopherol acetate can be hydrolyzed to d-alpha-tocopherol (VE) and absorbed in the small intestine. This product is intended for research use only.
  • Inhibits LPS-induced production of NO, prostaglandin E2 and pro-inflammatory cytokines
  • Exhibits slight suppression on the activation of NF-κB p50 and p65