Learn More
Medchemexpress LLC D-α-Tocopherol acetate | 58-95-7 | MFCD00072042 | 98.3% | 50 G
SDP
Catalog No. 5000412823
Encompass_Preferred
D-α-Tocopherol acetate can be hydrolyzed to d-alpha-tocopherol (VE) and absorbed in the small intestine. It has been shown to inhibit LPS-induced production of NO, prostaglandin E2, and pro-inflammatory cytokines (TNF-α, IFN-γ, IL-1β, and IL-6) in mouse peritoneal macrophages. Additionally, it exhibits a slight suppression on the activation of NF-κB p50 and p65.
  • Can be hydrolyzed to d-alpha-tocopherol (VE) and absorbed in the small intestine.
  • Inhibits LPS-induced production of NO, prostaglandin E2, and pro-inflammatory cytokines.
  • Slightly suppresses activation of NF-κB p50 and p65.
  • For research use only.
Catalog No. 50-004-12823 Supplier Medchemexpress LLC Supplier No. HYB127850G
D-α-Tocopherol acetate can be hydrolyzed to d-alpha-tocopherol (VE) and absorbed in the small intestine. It has been shown to inhibit LPS-induced production of NO, prostaglandin E2, and pro-inflammatory cytokines (TNF-α, IFN-γ, IL-1β, and IL-6) in mouse peritoneal macrophages. Additionally, it exhibits a slight suppression on the activation of NF-κB p50 and p65.
  • Can be hydrolyzed to d-alpha-tocopherol (VE) and absorbed in the small intestine.
  • Inhibits LPS-induced production of NO, prostaglandin E2, and pro-inflammatory cytokines.
  • Slightly suppresses activation of NF-κB p50 and p65.
  • For research use only.