Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-α-Tocopherol succinate, MilliporeSigma™ Supelco™

Catalog No. 111014200 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4200 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4200 Supplier MilliporeSigma Supelco Supplier No. 47782
Add to Cart
Add to Cart

D-α-Tocopherol succinate is a potent antioxidant tocopherol with antitumor activity and inhibits colon cancer liver metastases.

Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 4345-03-3
Physical Form Neat
Linear Formula C33H54O5
MDL Number MFCD00072055
Synonym Vitamin E succinate
Type Analytical Standard
Quantity 100 mg
Formula Weight 530.78
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.