Learn More
Medchemexpress LLC D-α-Tocopherylquinone | 7559-04-8 | 1 ML
SDP
Catalog No. 5000421180
Encompass_Preferred
D-α-Tocopherylquinone is a quinone isolated from Phaeodactylum tricornutum and an oxidation product of α-Tocopherol (vitamin E). It functions as an anticoagulant and antioxidant, reducing cellular oxidative damage caused by oxidized lipids.
  • Acts as an anticoagulant and antioxidant.
  • Reduces cellular oxidative damage caused by oxidized lipids.
  • Binds to a liver cytosolic protein with a molecular mass of about 40 kDa.
  • Binds to glutathione-S-transferase (GST) for transport or excretion.
  • Inhibits GST activity dose-dependently.
Catalog No. 50-004-21180 Supplier Medchemexpress LLC Supplier No. HYN285310MM/1ML
D-α-Tocopherylquinone is a quinone isolated from Phaeodactylum tricornutum and an oxidation product of α-Tocopherol (vitamin E). It functions as an anticoagulant and antioxidant, reducing cellular oxidative damage caused by oxidized lipids.
  • Acts as an anticoagulant and antioxidant.
  • Reduces cellular oxidative damage caused by oxidized lipids.
  • Binds to a liver cytosolic protein with a molecular mass of about 40 kDa.
  • Binds to glutathione-S-transferase (GST) for transport or excretion.
  • Inhibits GST activity dose-dependently.