Learn More
Medchemexpress LLC D-α-tocopherylquinone | 7559-04-8 | MFCD00135657 | 98.9% | 446.71 | C29H50O3 | 10 MG
SDP
Catalog No. 5000289425
Encompass_Preferred
  • Oxidation product of α-tocopherol with quinone structure.
  • Exhibits antioxidant and anticoagulant activity.
  • Reduces oxidative damage from oxidized lipids.
  • Interacts with glutathione-S-transferase (GST).
  • Used in reactive oxygen species (ROS) and lipid peroxidation research.
  • Molecular formula C29H50O3; MW 446.71 Da.
Catalog No. 50-002-89425 Supplier Medchemexpress LLC Supplier No. HYN285310MG
  • Oxidation product of α-tocopherol with quinone structure.
  • Exhibits antioxidant and anticoagulant activity.
  • Reduces oxidative damage from oxidized lipids.
  • Interacts with glutathione-S-transferase (GST).
  • Used in reactive oxygen species (ROS) and lipid peroxidation research.
  • Molecular formula C29H50O3; MW 446.71 Da.