Learn More
Glass Expansion D-Torch with Ceramic Outer Tube for Elan/NexION
SDP
Catalog No. 501907024
Encompass_Preferred
D-Torch with ceramic outer tube & installation guide. Compatible with PerkinElmer Elan, NexION 300/350 ICP-MS.A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable ceramic outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) HF (alumina) or high purity sapphire.The ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 31-808-2847, 501907158, 501907155.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7024 Supplier Glass Expansion Supplier No. 308082937
D-Torch with ceramic outer tube & installation guide. Compatible with PerkinElmer Elan, NexION 300/350 ICP-MS.A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable ceramic outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) HF (alumina) or high purity sapphire.The ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 31-808-2847, 501907158, 501907155.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001