Learn More
Glass Expansion D-Torch for Optima 8x00 (with Quartz Outer Tube)
SDP
Supplier:  Glass Expansion 308083300
Encompass_Preferred
D-Torch with quartz outer tube & installation guide, requires choice of injector. Compatible with PerkinElmer Optima 8000/8300 DV Series ICP-OES.A patented, precision-engineered, demountable torch, incorporating a ceramic intermediate tube for greater robustness & demountable outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). Compatible with all EMT injectors.The D-Torch is the only design with an optional, demountable ceramic outer tube (P/N 501907170). The quartz outer tube is ideal for most applications while the ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 31-808-3292, 501907204, 501907155Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7027