Learn More
Glass Expansion D-Torch PE AVIO 200
SDP
Supplier:  Glass Expansion 308083800
Encompass_Preferred
D-Torch with quartz outer tube & installation guide, compatible with PerkinElmer Avio 200/220 Series. Injector adaptor not included, compatible with PE design N0790141 & PE Avio injectors. A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable quartz outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). The D-Torch is the only design with an optional ceramic outer tube (P/N 501907262). The quartz outer tube is ideal for most applications while the ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 501907261, 31-808-2818, 501907259, 70-803-1576Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7038