Learn More
Glass Expansion D-Torch for Optima 2/4/5/7000 DV (with Quartz Outer Tube)
SDP
Catalog No. 501907022
Encompass_Preferred
D-Torch with quartz outer tube & installation guide, requires choice of injector. Compatible with PerkinElmer Optima 2/4/5/7000 DV ICP-OES.A patented, precision-engineered, demountable torch, incorporating a ceramic intermediate tube for greater robustness & demountable outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). Compatible with all EMT injectors.The D-Torch is the only design with an optional, demountable ceramic outer tube (P/N 501907212). The quartz outer tube is ideal for most applications while the ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 31-808-2981, 501907169, 501907155, 31-808-3377, 70-803-0762Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7022 Supplier Glass Expansion Supplier No. 308082926
D-Torch with quartz outer tube & installation guide, requires choice of injector. Compatible with PerkinElmer Optima 2/4/5/7000 DV ICP-OES.A patented, precision-engineered, demountable torch, incorporating a ceramic intermediate tube for greater robustness & demountable outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). Compatible with all EMT injectors.The D-Torch is the only design with an optional, demountable ceramic outer tube (P/N 501907212). The quartz outer tube is ideal for most applications while the ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 31-808-2981, 501907169, 501907155, 31-808-3377, 70-803-0762Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001