Learn More
Thermo Scientific Outer Tube for Radial D-Torch | Quartz | 1 EA
Supplier:  Thermo Scientific 842312052208
Encompass
Quartz Outer Tube for Radial D-Torch for iCAP 7000PRO Series ICP-OES systems. Resistant to chemical degradation, maintains properties at high temperatures, prevents devitrification associated with traditional quartz torches.
Catalog No. NC1368909