Learn More
Glass Expansion D-Torch & installation guide. Compatible with PerkinElmer 1000/2000 ICP-MS.A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable quartz outer tube that can be replaced.
SDP
Catalog No. 502703164
Encompass_Preferred
D-Torch & installation guide. Compatible with PerkinElmer 1000/2000 ICP-MS.A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable quartz outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). Compatible with all EMT injectors.The quartz outer tube is ideal for most aqueous applications. Replacement parts include 31-808-3928, 31-808-3942, 501907155.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3164 Supplier Glass Expansion Supplier No. 308083927
D-Torch & installation guide. Compatible with PerkinElmer 1000/2000 ICP-MS.A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable quartz outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). Compatible with all EMT injectors.The quartz outer tube is ideal for most aqueous applications. Replacement parts include 31-808-3928, 31-808-3942, 501907155.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001