Learn More
Glass Expansion D-Torch Pro Duo with 2.0mm Quartz Injector
Supplier: Glass Expansion 308084150
D-Torch complete assembly with 2.0mm ID quartz injector & installation guide. Compatible with Thermo PRO Duo ICP-OES.A patented, precision-engineered, demountable torch, incorporating a ceramic inner tube for greater robustness & demountable quartz outer tube that can be replaced when it fails rather than replacing the entire torch. D-Torch also features an interchangeable injector, allowing installation of a specific injector, whether it be for aqueous (~2.0mm ID), organics (<2.0mm ID), high TDS (>2.0mm ID) or HF (alumina). Compatible with all EMT injectors.The D-Torch is the only design with an optional ceramic outer tube (P/N NC2046972). The quartz outer tube is ideal for most applications while the ceramic outer tube is best for high TDS, organics, or HF matrices. Replacement parts include 31-808-4125, NC2046973, NC2046972, 50-169-4654, 31-808-2707, 31-808-2895, 31-808-2897, 70-803-1520.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001