Learn More
Medchemexpress LLC D-(+)-trehalose-13C12 | 1313730-07-2 | MFCD19704785 | 99.9% | 354.21 | 13C12H22O11 | 1mg
SDP
Catalog No. 5000269554
Encompass_Preferred
D-( )-Trehalose-13C12 is a 13C isotope labeled version of D-( )-Trehalose (HY-N1132) D-( )-Trehalose which is widespread can be used as a food ingredient and pharmaceutical excipient[1]
Catalog No. 50-002-69554 Supplier Medchemexpress LLC Supplier No. HYW7683451MG
D-( )-Trehalose-13C12 is a 13C isotope labeled version of D-( )-Trehalose (HY-N1132) D-( )-Trehalose which is widespread can be used as a food ingredient and pharmaceutical excipient[1]