Learn More
Medchemexpress LLC D-(+)-Trehalose | 99-20-7 | 342.30 | 1 ML
SDP
Catalog No. 5000415575
Encompass_Preferred
D-(+)-Trehalose (α,α-Trehalose) is an orally active disaccharide, showing anti-desiccant and cryopreservative activities. It acts as an osmolyte, a stress protectant, and helps in the storage and transport of carbon. D-(+)-Trehalose can be used as a food ingredient and pharmaceutical excipient.
  • Orally active disaccharide.
  • Shows anti-desiccant activity.
  • Exhibits cryopreservative activities.
  • Functions as an osmolyte.
  • Acts as a stress protectant.
  • Assists in carbon storage and transport.
  • Can be used as a food ingredient.
  • Suitable as a pharmaceutical excipient.
  • Is a microbial metabolite.
  • Is a human endogenous metabolite.
Catalog No. 50-004-15575 Supplier Medchemexpress LLC Supplier No. HYN113210MM/1ML
D-(+)-Trehalose (α,α-Trehalose) is an orally active disaccharide, showing anti-desiccant and cryopreservative activities. It acts as an osmolyte, a stress protectant, and helps in the storage and transport of carbon. D-(+)-Trehalose can be used as a food ingredient and pharmaceutical excipient.
  • Orally active disaccharide.
  • Shows anti-desiccant activity.
  • Exhibits cryopreservative activities.
  • Functions as an osmolyte.
  • Acts as a stress protectant.
  • Assists in carbon storage and transport.
  • Can be used as a food ingredient.
  • Suitable as a pharmaceutical excipient.
  • Is a microbial metabolite.
  • Is a human endogenous metabolite.