Learn More
Medchemexpress LLC D-(+)-Trehalose | 99-20-7 | 342.30 | 500 MG
SDP
Catalog No. 5000414635
Encompass_Preferred
D-(+)-Trehalose (α,α-Trehalose) is an orally active disaccharide. It acts as an osmolyte and stress protectant, helping in the storage and transport of carbon. It can be used as a food ingredient and pharmaceutical excipient.
  • Orally active disaccharide
  • Anti-desiccant and cryopreservative activities
  • Acts as an osmolyte
  • Stress protectant
  • Helps in storage and transport of carbon
  • Can be used as a food ingredient and pharmaceutical excipient
Catalog No. 50-004-14635 Supplier Medchemexpress LLC Supplier No. HYN1132A500MG
D-(+)-Trehalose (α,α-Trehalose) is an orally active disaccharide. It acts as an osmolyte and stress protectant, helping in the storage and transport of carbon. It can be used as a food ingredient and pharmaceutical excipient.
  • Orally active disaccharide
  • Anti-desiccant and cryopreservative activities
  • Acts as an osmolyte
  • Stress protectant
  • Helps in storage and transport of carbon
  • Can be used as a food ingredient and pharmaceutical excipient