Learn More
Medchemexpress LLC D-(+)-Trehalose | 99-20-7 | 342.30 | 500 MG
SDP
Catalog No. 5000415084
Encompass_Preferred
D-(+)-Trehalose is an orally active disaccharide that exhibits anti-desiccant and cryopreservative activities. It functions as an osmolyte and stress protectant, aiding in carbon storage and transport. It can be utilized as a food ingredient and a pharmaceutical excipient.
  • Orally active disaccharide
  • Exhibits anti-desiccant and cryopreservative activities
  • Functions as an osmolyte and stress protectant
  • Aids in carbon storage and transport
  • Can be utilized as a food ingredient and pharmaceutical excipient
  • No toxic effects on Chinese hamster ovary (CHO) cells at 5000 μg/mL
Catalog No. 50-004-15084 Supplier Medchemexpress LLC Supplier No. HYN1132500MG
D-(+)-Trehalose is an orally active disaccharide that exhibits anti-desiccant and cryopreservative activities. It functions as an osmolyte and stress protectant, aiding in carbon storage and transport. It can be utilized as a food ingredient and a pharmaceutical excipient.
  • Orally active disaccharide
  • Exhibits anti-desiccant and cryopreservative activities
  • Functions as an osmolyte and stress protectant
  • Aids in carbon storage and transport
  • Can be utilized as a food ingredient and pharmaceutical excipient
  • No toxic effects on Chinese hamster ovary (CHO) cells at 5000 μg/mL