Learn More
Medchemexpress LLC D-Trehalose, Carrier Protein | 99-20-7 | 99.8% | 50 MG
SDP
Catalog No. 5000409666
Encompass_Preferred
D-Trehalose is a disaccharide formed by a 1,1-glycosidic bond between two α-glucose units. It is widely used as a food ingredient and pharmaceutical excipient, known to protect and stabilize proteins. For long-term storage, recombinant protein solution should be diluted further with 5% D-Trehalose, Carrier Protein.
  • Stabilizes cell membranes and protein structures.
  • Resists desiccation.
  • Pharmaceutical excipient, used as an excipient.
  • Inhibits collagen degradation.
  • Forms a unique protective film on cell surfaces under harsh environmental conditions (high temperature, extreme cold, high osmotic pressure, desiccation).
  • Can be used as a carrier protein for long-term storage of recombinant proteins, preventing adherence to plastic tube walls and protecting protein activity.
Catalog No. 50-004-09666 Supplier Medchemexpress LLC Supplier No. HYCP00150MG
D-Trehalose is a disaccharide formed by a 1,1-glycosidic bond between two α-glucose units. It is widely used as a food ingredient and pharmaceutical excipient, known to protect and stabilize proteins. For long-term storage, recombinant protein solution should be diluted further with 5% D-Trehalose, Carrier Protein.
  • Stabilizes cell membranes and protein structures.
  • Resists desiccation.
  • Pharmaceutical excipient, used as an excipient.
  • Inhibits collagen degradation.
  • Forms a unique protective film on cell surfaces under harsh environmental conditions (high temperature, extreme cold, high osmotic pressure, desiccation).
  • Can be used as a carrier protein for long-term storage of recombinant proteins, preventing adherence to plastic tube walls and protecting protein activity.